Merck
  • In pursuit of drugs for American trypanosomiasis: evaluation of some "standards" in a mouse model.
In pursuit of drugs for American trypanosomiasis: evaluation of some "standards" in a mouse model.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.) (1997-12-24)
K E Kinnamon, B T Poon, W L Hanson, V B Waits
ABSTRACT
Forty-nine "standard" compounds known to be useful in the treatment of other diseases were tested for their suppressive activity against the trypomastigotes of Trypanosoma cruzi-infected mice. The most active was the antidepressant protriptyline, which was almost three times as effective as the reference drug, nifurtimox. A major value of the present data is to demonstrate the refractoriness of the T. cruzi parasite against many of the drug standards that have known biological activity.