Merck
  • Parathyroid hormone increases prostaglandin G/H synthase-2 transcription by a cyclic adenosine 3',5'-monophosphate-mediated pathway in murine osteoblastic MC3T3-E1 cells.
Parathyroid hormone increases prostaglandin G/H synthase-2 transcription by a cyclic adenosine 3',5'-monophosphate-mediated pathway in murine osteoblastic MC3T3-E1 cells.
Endocrinology (1997-09-01)
S Tetradis, C C Pilbeam, Y Liu, H R Herschman, B E Kream
ABSTRACT
PTH increased PG synthase-2 transcription in osteoblastic MC3T3-E1 cells at 30 min, as assessed by nuclear run-on assays. To determine the signaling pathways used by PTH, the activity of a construct containing the PG synthase-2 gene between nucleotides -963 and +70 linked to a luciferase reporter was analyzed in stably transfected MC3T3-E1 cells. Agents that activate the cAMP-protein kinase A or protein kinase C pathways increased PG synthase-2 promoter activity. In contrast, the calcium ionophore ionomycin was ineffective. The protein kinase A inhibitor H89 blocked PTH stimulation of PG synthase-2 promoter activity, whereas an overnight pre-incubation with phorbol ester to down-regulate protein kinase C did not. PTH-(3-34), a peptide that has greatly reduced ability to activate the cAMP-protein kinase A pathway, did not increase PG synthase-2 transcription or promoter activity. PTH could induce PG synthase-2 messenger RNA accumulation and PG synthase-2 transcription in the presence of cycloheximide. In addition, PTH-stimulated PG synthase-2 transcription was maintained at a high level at 2 h in the presence of cycloheximide. We conclude that PTH rapidly increases PG synthase-2 transcription in MC3T3-E1 cells, mainly through a cAMP-protein kinase A-mediated pathway without the need for protein synthesis. In contrast, the attenuation of increased PG synthase-2 transcription by PTH requires de novo protein synthesis.
Sigma-Aldrich
Parathyroid Hormone Fragment 1-34 bovine, ≥97% (HPLC), powder