Merck
Comparative evaluation of determination of beta-lactam intermediates.
Hindustan antibiotics bulletin (1993-08-01)
B S Deshpande, S S Ambedkar, J G Shewale
ABSTRACT
The Schiff's base formation between 6-aminopenicillanic acid, 7-aminodesacetoxycephalosporanic acid and 7-aminocephalosporanic acid and p-dimethylaminobenzaldehyde (PDAB) was investigated. The factors that affect the reaction such as concentration of PDAB, time and pH were studied and optimised for estimation of these intermediates.
Product Description
Supelco
4-(Dimethylamino)benzaldehyde solution, 10 g/L in isopropanol, for TLC derivatization
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, puriss. p.a., reag. Ph. Eur., ≥99% (perchloric acid titration)
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, ACS reagent, 99%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, 98%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
Millipore
Kovac′s reagent for indoles, suitable for microbiology
Millipore
Kovac′s reagent for indoles, suitable for microbiology