Merck
  • Synthesis of polymer-bound 6-thiolatomercury and 6-mercuric sulfonate DOPA precursors and their halodemercuration reactivity.
Synthesis of polymer-bound 6-thiolatomercury and 6-mercuric sulfonate DOPA precursors and their halodemercuration reactivity.
Nuclear medicine and biology (1995-01-01)
K Kawai, M A Channing, D O Kiesewetter, W C Eckelman
ABSTRACT
Fluorodemercuration has the greatest utility for the preparation of 6-[18F]DOPA, but requires separation from unreacted mercury precursor and other mercury-containing compounds. One approach is the development of a polymer-bound mercury precursor. In this study, polymer-bound 6-thiolatomercury and 6-mercuric sulfonate DOPA derivatives, and its monomeric analogs were synthesized. Fluorodemercuration of monomeric analog of mercuric sulfonate gave half the yield (14-15%) while iododemercuration gave the same yield (38%) compared with a 6-mercuric trifluoroacetate protected DOPA. The mercuric sulfonate undergoes halodemercuration, so polymer-bound halodemercuration precursors may be useful as precursors of 6-[18F]DOPA.
Sigma-Aldrich
Mercury(II) sulfate, puriss., ≥98%
Supelco
Mercury(II) sulfate, ACS reagent, for preparation of solution for COD determination according to DIN 38409, part 41, ≥99%