Merck
  • [Synthesis and structural study of 5-arylidene thiazolidine-2,4-diones and 3-substituted-4-thio-imidazolidine-2-ones].
[Synthesis and structural study of 5-arylidene thiazolidine-2,4-diones and 3-substituted-4-thio-imidazolidine-2-ones].
Annales pharmaceutiques francaises (1995-01-01)
J F Albuquerque, L C Azevedo, S L Galdino, J Chantegrel, I R Pitta, C Luu-Duc
ABSTRACT
The synthesis and the physico-chemical properties of four 3-(4-bromophenacyl)-5-arylidene-thiazolidine-2,4-diones, two 3-(4-bromobenzyl)-5-arylidene-thiazolidine-2,4-diones and seven 3-(4-chlorobenzyl)-5-arylidene-4-thio-imidazolidine-2-ones were described. These products were synthetized by the aldolisation-crotonisation reaction between aromatic aldehydes and substituted thiazolidinediones or thio-imidazolidinones.
Sigma-Aldrich
2-Imidazolidone, 96%