Merck
Mycotoxin production by Chaetomium spp. and related fungi.
Canadian journal of microbiology (1981-08-01)
S Sekita, K Yoshihira, S Natori, S Udagawa, T Muroi, Y Sugiyama, H Kurata, M Umeda
ABSTRACT
Screening for mycotoxin production by Chaetomium spp. and related fungi on rice culture was conducted by a combination of cytotoxicity tests using HeLa cells and thin-layer chromatography. Producers of sterigmatocystin, O-methylsterigmatocystin, chaetochromin, chaetocin, chetomin, cochliodinols, and mollicellin G were found and the taxonomic significance of these findings is discussed.
Sigma-Aldrich
Chetomin, from Chaetomium cochliodes, ≥98% (HPLC)