Merck
  • Correction for contaminant fluorescence in frequency-domain fluorometry.
Correction for contaminant fluorescence in frequency-domain fluorometry.
Analytical biochemistry (1987-02-01)
J R Lakowicz, R Jayaweera, N Joshi, I Gryczynski
ABSTRACT
We describe a general method to correct for contaminant fluorescence when using the technique of frequency-domain fluorometry. The method can be applied regardless of the origin of the background signal, from scattered light, impurity fluorescence, or both. The procedure requires measurement of the frequency-dependent phase and modulation of the background at enough frequencies to approximate the decay law of the background. We also describe a general method to propagate the uncertainties in the measured phase and modulation values into the corrected values. This propagation is necessary to ensure proper weighting of the frequency-dependent data in the least-squares fitting algorithms. The practical usefulness of this correction method is demonstrated using frequency-domain data for one and two component mixtures which were deliberately contaminated with scattered light and/or other fluorophores.
Sigma-Aldrich
1,4-Bis(5-phenyl-2-oxazolyl)benzene, BioReagent, suitable for scintillation
Supelco
9,10-Diphenylanthracene, analytical standard
Sigma-Aldrich
9,10-Diphenylanthracene, 97%