Merck
  • The promoting effects of food dyes, erythrosine (Red 3) and rose bengal B (Red 105), on thyroid tumors in partially thyroidectomized N-bis(2-hydroxypropyl)-nitrosamine-treated rats.
The promoting effects of food dyes, erythrosine (Red 3) and rose bengal B (Red 105), on thyroid tumors in partially thyroidectomized N-bis(2-hydroxypropyl)-nitrosamine-treated rats.
Japanese journal of cancer research : Gann (1988-03-01)
Y Hiasa, M Ohshima, Y Kitahori, N Konishi, T Shimoyama, Y Sakaguchi, H Hashimoto, S Minami, Y Kato
ABSTRACT
The effects of erythrosine (Red 3), rose bengal B (Red 105) and thyroidectomy on the development of thyroid tumor were examined in male Wistar rats treated with N-bis(2-hydroxypropyl)nitrosamine (DHPN). Red 3 and Red 105 were used at 4% in the basal diet and were administered for 19 weeks from week 2 to 20. Thyroidectomy was performed by resection of the left lobe at week 4. Single injection of DHPN was performed intraperitoneally at 280 mg per 100 g body weight at the beginning of the experiment. Red 3 and Red 105 significantly promoted the development of thyroid tumors in thyroidectomized rats given DHPN, but had no significant effect in non-thyroidectomized rats. The incidence of thyroid tumors was 91% in rats with partial thyroidectomy, Red 3 and DHPN, 100% in rats with partial thyroidectomy, Red 105 and DHPN, and 64% in rats with partial thyroidectomy and DHPN. Serum TSH was 5.5 +/- 3.1 ng/ml in rats with partial thyroidectomy, Red 3 and DHPN, 2.1 +/- 2.2 ng/ml in rats with partial thyroidectomy, Red 105 and DHPN, and 1.5 +/- 0.5 ng/ml in rats with partial thyroidectomy and DHPN.
Sigma-Aldrich
Rose Bengal lactone, 95%
Sigma-Aldrich
Rose bengal, certified by the Biological Stain Commission, Dye content ≥80 %
Sigma-Aldrich
Rose bengal, Dye content 95 %, ≥96% (HPLC)