Merck
Evaluation of the teratogenic potential of octyl acetate in rats.
Fundamental and applied toxicology : official journal of the Society of Toxicology (1989-08-01)
W C Daughtrey, P J Wier, K A Traul, R W Biles, G F Egan
ABSTRACT
Octyl acetate (CAS RN 108419-32-5) was administered via oral gavage to pregnant Sprague-Dawley rats on Gestation Days 6 through 15 at dose levels of 0, 0.1, 0.5, and 1.0 g/kg. The dams were weighed and observed for clinical signs of toxicity during pregnancy, and food consumption was measured. On Gestation Day 20 the dams were sacrificed and the fetuses were examined for external, visceral, and skeletal malformations and variations. The mid- and high-dose levels resulted in maternal toxicity as evidenced by reductions in body weight gain and food consumption. There were no statistically significant effects on embryo-fetal lethality or fetal growth for any treatment group. The number of litters with at least one malformed fetus and the mean percentage of the litter malformed were significantly (p less than 0.05) elevated in the high-dose group only. The results of the present study demonstrate that octyl acetate produced some evidence of developmental toxicity at a dose (1.0 g/kg) that was maternally toxic. Developmental toxicity was not observed at the maternally toxic 0.5 g/kg dose level or the maternally nontoxic dose level (0.1 g/kg). Therefore, these data indicate that octyl acetate is not a selective developmental toxicant in the rat.
Sigma-Aldrich
Octyl acetate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Octyl acetate, ≥98%, FCC, FG
Sigma-Aldrich
Octyl acetate, ≥99%