Merck
  • A phosphorbisamidite approach to the synthesis of oligodeoxyribonucleotides and their analogues.
A phosphorbisamidite approach to the synthesis of oligodeoxyribonucleotides and their analogues.
Nucleic acids symposium series (1989-01-01)
K Yamana, Y Nishijima, H Nakano, O Sangen, H Ozaki, T Shimidzu
ABSTRACT
Utilities of deoxyribonucleoside 3'-O-phosphorbisdiethylamidites in the synthesis of oligodeoxyribonucleotides and their analogues are described.
Sigma-Aldrich
Tris(diethylamino)phosphine, 97%