Merck
  • Cyclodextrin-based combinatorial polymers: efficient binders of pharmaceuticals in water.
Cyclodextrin-based combinatorial polymers: efficient binders of pharmaceuticals in water.
Chimia (2013-08-16)
Patrick Shahgaldian, Philippe F-X Corvini
ABSTRACT
Cyclodextrins are cyclic oligomers of glucose; they are widely used in a large range of industrial applications because of their molecular inclusion properties. We used cyclodextrins to prepare different libraries of polymers and tested their ability to selectively recognize pharmaceuticals in water. It was demonstrated that the chemical composition of the polymer strongly influences its binding properties. The developed strategy can be used to produce selective sorbent nanomaterials of pharmaceuticals.
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Supelco
Cellulose, acid washed, powder, for column chromatography
Supelco
Cellulose, powder, for column chromatography
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture