Merck
  • Rapid degradation of hexachlorobenzene by micron Ag/Fe bimetal particles.
Rapid degradation of hexachlorobenzene by micron Ag/Fe bimetal particles.
Journal of environmental sciences (China) (2013-08-08)
Xiaoqin Nie, Jianguo Liu, Xianwei Zeng, Dongbei Yue
ABSTRACT
The feasibility of the rapid degradation of hexachlorobenzene (HCB) by micron-size silver (Ag)/iron (Fe) particles was investigated. Ag/Fe particles with different ratios (0, 0.05%, 0.09%, 0.20%, and 0.45%) were prepared by electroless silver plating on 300 mesh Fe powder, and were used to degrade HCB at different pH values and temperatures. The dechlorination ability of Fe greatly increased with small Ag addition, whereas too much added Ag would cover the Fe surface and reduce the effective reaction surface, thereby decreasing the extent of dechlorination. The optimal Ag/Fe ratio was 0.09%. Tafel polarization curves showed that HCB was rapidly degraded at neutral or acidic pH, whereas low pH levels severely intensified H2 production, which consumed the reducing electrons needed for the HCB degradation. HCB degradation was more sensitive to temperature than pH. The rate constant of HCB dechlorination was 0.452 min- at 85 degrees C, 50 times higher than that at 31 degrees C. HCB was degraded in a successive dechlorination pathway, yielding the main products 1,2,4,5-tetrachlorobenzene and 1,2,4-trichlorobenzene within 2 hr.
Product Description
Supelco
Hexachlorobenzene solution, certified reference material, 1000 μg/mL in acetone
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Supelco
Hexachlorobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, granular, >250 μm, 99.99% trace metals basis