Merck
  • An efficient heterogeneous Cu-grafted mesoporous organosilicas nanocatalyst for two and three component C-S coupling reactions.
An efficient heterogeneous Cu-grafted mesoporous organosilicas nanocatalyst for two and three component C-S coupling reactions.
Journal of nanoscience and nanotechnology (2013-08-02)
John Mondal, Noor Salam, Asim Bhaumik
ABSTRACT
Furfural-imine functionalized mesoporous organosilica material has been synthesized by post-synthesis surface functionalization of 2D-hexagnoal mesoporous SBA-15 with organosilane precursor 3-aminopropyltriethoxy-silane (APTES) followed by Schiff-base condensation with furfural. On the other hand, furfural-imine functionalized MCM-41 has been synthesized by Schiff-base condensation of furfural and 3-aminopropyltriethoxy-silane (APTES) followed by its hydrothermal co-condensation with tetraethylorthosilicate (TEOS) in the presence of a cationic surfactant CTAB. Subsequent reaction of the imine-functionalized mesoporous organosilicas with Cu(OAc)2 in absolute ethanol produced Cu(II)-grafted mesoporous nanocatalysts 1 and 2, respectively. Powder XRD, HR TEM, FE SEM, N2 sorption and EPR experimental tolls are employed to characterize these Cu-grafted furfural-imine functionalized nanocatalysts. Cu-grafted MCM-41 (1) showed very good catalytic efficiency for the coupling of aryl bromides and thiophenol under aerobic conditions to produce different thioethers. On the other hand, Cu-anchored mesoporous SBA-15 nanocatalyst (2) exhibited high catalytic activity for one-pot three component coupling of different aryl halides with thiourea and benzyl bromide in aqueous medium to produce different aryl alkyl thioethers in very good yields. Both Cu-grafted mesoporous nanocatalysts can be efficiently reused for several reaction cycles.
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, ≥99.9% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Silica
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Copper (S, P), BCR®, certified reference material, chips
Copper, IRMM®, certified reference material, 0.5 mm wire
Copper, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Copper, powder, 99.999% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O