Merck
  • Enhancement of biocompatibility of metal implants by nanoscale tiN/NbN multilayer coatings.
Enhancement of biocompatibility of metal implants by nanoscale tiN/NbN multilayer coatings.
Journal of nanoscience and nanotechnology (2013-08-02)
B Subramanian
ABSTRACT
Titanium nitride (TiN)/niobium nitride (NbN) nanostructured multilayer coatings were prepared by DC reactive magnetron sputtering method using the combination of a titanium and niobium target and an Ar-N2 mixture discharge gas on to 316L stainless steel substrates. The coatings showed a polycrystalline structure with (111) for TiN and (101) for NbN preferential growth. Raman spectroscopy measurements on the multilayer films exhibited the characteristic peaks at 212, 303, 458 and 578 cm-1. A higher hardness of 38 GPa was observed for TiN/NbN coatings. Electrochemical polarization tests were performed in simulated biological fluid solutions at 37 degreesC in order to determine and compare the corrosion behavior of the coated and uncoated 316L SS substrates. The TiN/NbN multilayer coatings could improve the corrosion resistance of 316L SS substrate. The bacterial culture experiments were performed and the bacteria treated samples were examined by epi fluorescence microscope measurements.
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Niobium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.1 mm foil
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Niobium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm foil
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium nitride, <3 μm