Merck
  • Kinetics of enzymatic modification of quercetin with cysteine by horseradish peroxidase.
Kinetics of enzymatic modification of quercetin with cysteine by horseradish peroxidase.
Indian journal of biochemistry & biophysics (2013-08-01)
Sasa R Savic, Zivomir B Petronijevic
ABSTRACT
The kinetic mechanism of enzymatic modification of flavonol quercetin with L-cysteine by horseradish peroxidase (HRP) was studied. Reaction of modification of quercetin was followed by recording spectral changes over time at 380 nm. All reactions were performed in 100 mM phosphate buffer pH, 6.0 at 20 degrees C. Kinetic parameters were determined from graphics of linear Michaelis-Menten equation. The values obtained at specified intervals were: V(max) = 0.17 divided by 0.91 deltaA380/min, K(m) = 0.023 divided by 0.5 mM, k(cat) = 0.21 divided by 1.14 deltaA380/min x nM(-1) and V(max)/K(m) = 0.83 divided by 26.55 deltaA380/min x mM(-1). It was found that all investigated reactions of the modification of quercetin with L-cysteine by HRP followed an ordered mechanism. We propose that HRP initially reacts with H2O2 than with quercetin and finally with L-cysteine, leading to the introduction of L-cysteine in the structure of quercetin.
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, Vetec, reagent grade, 97%
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, Wacker Chemie AG, 98.5-101.0%
Supelco
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Cysteine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, from non-animal source, suitable for cell culture, meets EP, USP testing specifications