Merck
  • Platelet reactivity and clinical outcomes after coronary artery implantation of drug-eluting stents (ADAPT-DES): a prospective multicentre registry study.
Platelet reactivity and clinical outcomes after coronary artery implantation of drug-eluting stents (ADAPT-DES): a prospective multicentre registry study.
Lancet (London, England) (2013-07-31)
Gregg W Stone, Bernhard Witzenbichler, Giora Weisz, Michael J Rinaldi, Franz-Josef Neumann, D Christopher Metzger, Timothy D Henry, David A Cox, Peter L Duffy, Ernest Mazzaferri, Paul A Gurbel, Ke Xu, Helen Parise, Ajay J Kirtane, Bruce R Brodie, Roxana Mehran, Thomas D Stuckey
ABSTRACT
The relation between platelet reactivity and stent thrombosis, major bleeding, and other adverse events after coronary artery implantation of drug-eluting stents has been incompletely characterised. We aimed to determine the relation between platelet reactivity during dual therapy with aspirin and clopidogrel and clinical outcomes after successful coronary drug-eluting stent implantation. ADAPT-DES was a prospective, multicentre registry of patients successfully treated with one or more drug-eluting stents and given aspirin and clopidogrel at 10-15 US and European hospitals. We assessed platelet reactivity in those patients after successful percutaneous coronary intervention using VerifyNow point-of-care assays, and assigned different cutoffs to define high platelet reactivity. The primary endpoint was definite or probable stent thrombosis; other endpoints were all-cause mortality, myocardial infarction, and clinically relevant bleeding. We did a propensity-adjusted multivariable analysis to determine the relation between platelet reactivity and subsequent adverse events. This study is registered with ClinicalTrials.gov, number NCT00638794. Between Jan 7, 2008, and Sept 16, 2010, 8665 patients were prospectively enrolled at 11 sites, of which 8583 were eligible. At 1-year follow-up, stent thrombosis had occurred in 70 (0·8%) patients, myocardial infarction in 269 (3·1%), clinically relevant bleeding in 531 (6·2%), and death in 161 (1·9%) patients. High platelet reactivity on clopidogrel was strongly related to stent thrombosis (adjusted HR 2·49 [95% CI 1·43-4·31], p=0·001) and myocardial infarction (adjusted HR 1·42 [1·09-1·86], p=0·01), was inversely related to bleeding (adjusted HR 0·73 [0·61-0·89], p=0·002), but was not related to mortality (adjusted HR 1·20 [0·85-1·70], p=0·30). High platelet reactivity on aspirin was not significantly associated with stent thrombosis (adjusted HR 1·46 [0·58-3·64], p=0·42), myocardial infarction, or death, but was inversely related to bleeding (adjusted HR 0·65 [0·43-0·99], p=0·04). The findings from this study emphasise the counter-balancing effects of haemorrhagic and ischaemic complications after stent implantation, and suggest that safer drugs or tailored strategies for the use of more potent agents must be developed if the benefits of greater platelet inhibition in patients with cardiovascular disease are to be realised. Boston Scientific, Abbott Vascular, Medtronic, Cordis, Biosensors, The Medicines Company, Daiichi-Sankyo, Eli Lilly, Volcano, and Accumetrics.
Product Description
Ticlopidine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Ticlopidine hydrochloride, analytical standard, for drug analysis
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material