Merck
Synthesis of highly ordered mesoporous Co3O4 for gas sensing.
Journal of nanoscience and nanotechnology (2013-07-19)
D W Su, H Liu, H J Ahn, G X Wang
ABSTRACT
Highly ordered mesoporous Co3O4 nanostructures were prepared using SBA-15 silica as hard templates. The mesoporous structures were characterized by X-ray diffraction, high resolution transmission electron microscopy, and N2 adsorption/desorption isotherm analysis. The results demonstrated that the as-prepared mesoporous Co3O4 has an ordered P6mm symmetric mesoporous structure. The optical absorption properties of the mesoporous Co3O4 were investigatted by UV-Vis spectroscopy and the results indicate that the mesoporous Co3O4 materials are semiconducting with direct band gaps of 2, 1.385 and 0.38 eV. The gas-sensing performance of the mesoporous Co3O4 was tested towards a series of typical solvents. They demonstrated a good sensing performance towards these vapour with rapid response and high sensitivity at low operating temperature.
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis