Merck
  • [Genes for cellulose-degradation and their expression conditions in Chaetomium globosum NK102].
[Genes for cellulose-degradation and their expression conditions in Chaetomium globosum NK102].
Wei sheng wu xue bao = Acta microbiologica Sinica (2013-07-19)
Yang Hu, Jiannan Bi, Xiaoran Hao, Huang Chen, Jiao Pan, Xudong Zhu
ABSTRACT
Use of renewable plant biomass is an active area in current biotechnology research. This report explores the cellulose-degradation system and the factors affecting cellulase gene expression in Chaetomium globosum NK102. In the sequenced genome, we identified 10 cellulase genes by sequence homology alignment. We used a high-throughput sequencing technology (RNA-Seq) to monitor the differential expression of the genes under different culture conditions. We observed that cellulase activity increased with time in the fungal cultures. Accordingly, transcription of the genes encoding cellobiohydrolase, cellobiose dehydrogenase and endoglucanase (cbh1, cdh and eglI) was higher than the others. Expression of the transcriptional repressors, ACE I and CreA, decreased with the time, whereas expression of Hap2/3/5 complex was upregulated. In different carbon sources, cellulase activity and their gene transcription were repressed by glucose and were activated by cellobiose. Sorbitol had no significant effect. Interestingly, light affected positively the expression of these cellulase genes. Differential RNA-seq analysis can make preliminary exploration on the expression regulation of cellulase genes. Expression of the genes in C. globosum was determined by different culture conditions. This study has shown a molecular system of cellulose-degradation in C. globosum and provides information for interpretation of carbohydrate metabolism.
Product Description
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, powder, suitable for column chromatography