Merck
  • [Autochthonous probiotics in prevention of infectious and inflammatory diseases of a human in the altered habitats].
[Autochthonous probiotics in prevention of infectious and inflammatory diseases of a human in the altered habitats].
Vestnik Rossiiskoi akademii meditsinskikh nauk (2013-07-04)
V K Il'in, A N Suvorov, N V Kiriukhina, N A Usanova, L V Starkova, V V Boiarintsev, A B Karaseva
ABSTRACT
Dysbiotic shifts in intestinal and pharyngeal microflora were studied in 22 normal volunteers in 9-, 14-, 105- and 520-d chamber experiments simulating some of the spaceflight factors. Two preparations were administered to prevent pharyngeal and intestinal dysbiosis: oral dry probiotic based on indigenous intestinal Enterococci and topical collagen-immobilized Lactobacterin based on indigenous intestinal Lactobacilli. Topical autopmrobiotic lactobacterin reduced the growth of opportunistic pathogens in the throat during the experiments. Oral autoprobiotic based on Enterococci reduced the content of intestinal opportunistic pathogens, supporting the high level of protecting microflora. Most of autostrains are free from pathogenicity factors, nevertheless, implementation of genetic testing of indigenous strains are reasonable.
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder