Merck
  • Effect of short peptides containing lysine and epsilon-aminocaproic acid on fibrinolytic activity of plasmin and topoisomerase II action on supercoiled DNA.
Effect of short peptides containing lysine and epsilon-aminocaproic acid on fibrinolytic activity of plasmin and topoisomerase II action on supercoiled DNA.
Acta poloniae pharmaceutica (2013-06-14)
Krystyna Midura-Nowaczek, Maciej Purwin, Agnieszka Markowska, Danuta Drozdowska, Magdalena Bruzgo
ABSTRACT
Effects of eight short peptides containing lysine and epsilon-aminocaproic acid (EACA) on prolongation of the clot lysis time, as well as hemolytic and antibacterial activities were investigated. Interaction with plasmids pBR322 and pUC19 with the use of ethidium bromide assay and determination of influence on the activity of topoisomerase I and II were also tested. Examined compounds inhibited fibrinolytic activity of plasmin and five of them were more active than EACA. Amides of dipeptides were most active antifibrinolytics (IC50 < 0.2 mM). According to the obtained data, the significant inhibition of fibrinolytic activity of plasmin was not associated with hemolytic effects. Examined compounds did not show antibacterial activity (MIC > 512 mg/L). DNA binding effects determined with the use of ethidium bromide were weak for all peptides and similar to those observed with EACA. Six compounds inhibited topoisomerase II action on supercoiled DNA.
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
6-Aminocaproic acid, BioUltra, ≥99%
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)
Supelco
Aminocaproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material