Merck
  • The effect of polaprezinc on gastric mucosal protection in rats with ethanol-induced gastric mucosal damage: comparison study with rebamipide.
The effect of polaprezinc on gastric mucosal protection in rats with ethanol-induced gastric mucosal damage: comparison study with rebamipide.
Life sciences (2013-06-08)
Hyuk Soon Choi, Ji-Youn Lim, Hoon Jai Chun, Min Lee, Eun Sun Kim, Bora Keum, Yeon Seok Seo, Yoon-Tae Jeen, Soon Ho Um, Hong Sik Lee, Chang Duck Kim, Ho Sang Ryu, Donggeun Sul
ABSTRACT
Polaprezinc (PZ), which consists of l-carnosine and zinc, is widely used to treat gastric ulcers. We compared the effects of PZ with those of rebamipide (RM) on the expression of inflammatory cytokines, antioxidants, growth factors, and heat shock proteins (HSP) in a rat model. Seventy Sprague-Dawley rats were randomly assigned to test groups according to the dose of PZ at 5, 10, or 30 mg/kg or RM at 10, 30, or 100 mg/kg. Next, we obtained ulcer indices from rats with ethanol-induced gastric mucosal damage. Western blot analysis was used to evaluate the expression of various target proteins. Pathological ulcer indices in the PZ and RM groups were significantly lower than those in the control group. The levels of inflammatory cytokines (interleukin 1β [IL-1β], IL-6, IL-8, and tumor necrosis factor α) decreased, whereas the levels of platelet-derived growth factor-B, vascular endothelial growth factor, and nerve growth factor significantly increased after PZ administration. Furthermore, the expression of antioxidants (superoxide dismutase 1 [SOD-1], SOD-2, heme oxygenase-1, glutathione S-transferase, peroxidredoxin-1, and peroxidredoxin-5) was significantly higher in the PZ group, and the levels of HSP 90, 70, 60, 47, 27, and 10 significantly increased with an increase in PZ dose. In a rat model of ethanol-induced gastric mucosal damage, PZ administration ameliorated ethanol-induced mucosal injury and showed protective effects on the mucosa by reducing the levels of inflammatory cytokines and increasing the expression of antioxidant enzymes and growth factors. Furthermore, PZ showed cytoprotective effects by increasing the HSP levels.
Sigma-Aldrich
Reagent Alcohol, reagent grade
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Alanine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Reagent Alcohol, suitable for HPLC
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
L-Carnosine, ~99%, crystalline
Sigma-Aldrich
Glutathione S-Transferase from equine liver, lyophilized powder, ≥25 units/mg protein
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology