Merck
  • Ultrasound-mediated transscleral delivery of macromolecules to the posterior segment of rabbit eye in vivo.
Ultrasound-mediated transscleral delivery of macromolecules to the posterior segment of rabbit eye in vivo.
Investigative ophthalmology & visual science (2013-06-01)
Wai-Leung Langston Suen, Hoi Sang Wong, Yu Yu, Laurence Chi Ming Lau, Amy Cheuk-Yin Lo, Ying Chau
ABSTRACT
This study aims to determine the in vivo effectiveness of low-frequency ultrasound in mediating the transport of macromolecules to the posterior segment of the eye via transscleral route. It investigates if damage is caused by ultrasound at the tested operation parameters on the posterior ocular tissues and visual function. Ultrasound (I(SATA) = 0.12 W/cm(2), center frequency = 40 kHz, 90-second continuous wave) was applied on the sclera of New Zealand white rabbits for one to three cycles. Solution of fluorescent dextran (70 kDa) was placed above sclera during and after ultrasound application to assess transscleral transport of macromolecules. Amount of dextran delivered to vitreous was determined by detection of fluorescence. Visual function of ultrasound-treated rabbits was examined by full-field electroretinography (ffERG). The effect of ultrasound on ocular tissue structures was examined by binocular indirect ophthalmoscope (BIO) and histology. Repeated ultrasound resulted in increasing concentration of dextran, which was otherwise undetectable in the vitreous. Transscleral barrier against dextran transport was restored to original value at 2 weeks postultrasound treatment. Studies from ffERG suggested that electric responses from neural transmission of retinal cells are normal at 1 day, 7 days, and 14 days after ultrasound applications. BIO and histology revealed no structural abnormality in posterior ocular tissues after ultrasound treatment. Low-frequency ultrasound significantly enhanced the penetration of macromolecules via transscleral route. No undesirable side effects have been found for up to 2 weeks after ultrasound application. The study supports that sonication is a potentially safe and effective method to modulate transscleral barriers for delivering macromolecular therapeutics to posterior segment of the eye.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Supelco
Dextran, analytical standard, for GPC, 150,000
Supelco
Dextran, analytical standard, for GPC, 12,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
Supelco
Dextran, analytical standard, for GPC, 670,000
Supelco
Dextran, analytical standard, for GPC, 1,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Supelco
Dextran, analytical standard, for GPC, 5,000
Supelco
Dextran, analytical standard, for GPC, 50,000