Merck
  • CD33 Alzheimer's disease locus: altered monocyte function and amyloid biology.
CD33 Alzheimer's disease locus: altered monocyte function and amyloid biology.
Nature neuroscience (2013-05-28)
Elizabeth M Bradshaw, Lori B Chibnik, Brendan T Keenan, Linda Ottoboni, Towfique Raj, Anna Tang, Laura L Rosenkrantz, Selina Imboywa, Michelle Lee, Alina Von Korff, Martha C Morris, Denis A Evans, Keith Johnson, Reisa A Sperling, Julie A Schneider, David A Bennett, Philip L De Jager
ABSTRACT
In our functional dissection of the CD33 Alzheimer's disease susceptibility locus, we found that the rs3865444(C) risk allele was associated with greater cell surface expression of CD33 in the monocytes (t50 = 10.06, P(joint) = 1.3 × 10(-13)) of young and older individuals. It was also associated with diminished internalization of amyloid-β 42 peptide, accumulation of neuritic amyloid pathology and fibrillar amyloid on in vivo imaging, and increased numbers of activated human microglia.
Product Description
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Supelco
Dextran, analytical standard, for GPC, 5,000
Supelco
Dextran, analytical standard, for GPC, 1,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Supelco
Dextran, analytical standard, for GPC, 50,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, for GPC, 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Supelco
Dextran, analytical standard, for GPC, 12,000
Sigma-Aldrich
Dextran, enzymatic synth.
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
Supelco
Dextran, analytical standard, for GPC, 150,000
Supelco
Dextran, analytical standard, for GPC, 80,000