Merck
  • Recurrent laryngeal nerve transection and anastomosis: rat laryngeal motoneuron survival and effect of the anastomosis site.
Recurrent laryngeal nerve transection and anastomosis: rat laryngeal motoneuron survival and effect of the anastomosis site.
The Annals of otology, rhinology, and laryngology (2013-05-24)
Michael J Pitman, Craig Berzofsky, Opeyemi Alli, Sansar Sharma
ABSTRACT
We investigated the quantity of recurrent laryngeal nerve motoneurons (RLNMs) that survive after transection and anastomosis of the rat recurrent laryngeal nerve (RLN), as well as the impact of the anastomosis site on RLN regeneration. Ten rats underwent right RLN transection and anastomosis. After 16 weeks, Fluoro-Ruby (FR) was applied to the RLN that was transected proximal or distal to the anastomosis site. The brain stems were harvested, and the nucleus ambiguus was evaluated for labeled RLNMs. The RLNM counts were compared to each other and to those from 3 control rats in which FR was applied to an acutely transected RLN. The number of RLNMs that were stained after RLN transection, anastomosis, and regeneration was consistent with the total number of RLNMs in the nucleus ambiguus of control rats. This finding confirms that most RLNMs survived after RLN transection and anastomosis. The quantity of labeled RLNMs was statistically similar whether the FR was applied proximal or distal to the anastomosis, implying that most of the viable axons that were present proximal to the anastomosis crossed into the distal nerve. Rat RLNMs survive nerve transection, anastomosis, and regeneration. The anastomosis site does not significantly impede axonal regeneration, and most of the axons traverse the anastomosis into the distal nerve.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Supelco
Dextran, analytical standard, for GPC, 25,000
Supelco
Dextran, analytical standard, for GPC, 670,000
Supelco
Dextran, analytical standard, for GPC, 12,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Supelco
Dextran, analytical standard, for GPC, 50,000
Supelco
Dextran, analytical standard, for GPC, 1,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Supelco
Dextran, analytical standard, for GPC, 80,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000