Merck
  • Magnesium sulfate maintenance infusion in women with preeclampsia: a randomized comparison between 2 gram per hour and 1 gram per hour.
Magnesium sulfate maintenance infusion in women with preeclampsia: a randomized comparison between 2 gram per hour and 1 gram per hour.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet (2013-05-23)
Dhirapatara Charoenvidhya, Saknan Manotaya
ABSTRACT
To compare the percentages of pregnant women with preeclampsia who reached the therapeutic serum magnesium levels between those who received maintenance magnesium sulfate infusion of 2 g/hour versus I g/hour Sixty women diagnosed of preeclampsia and magnesium sulfate that were considered for seizure prophylaxis were randomized into two groups. A loading dose of 5 g magnesium sulfate was given intravenously over 20 minutes to both groups. Maintenance doses of magnesium sulfate of 2 g/hour and 1 g/hour were given to the study and control groups, respectively. The maintenance dose was continued until 24 hours postpartum. Blood samples for serum magnesium were collected at 0, 1/2, 2, and 4 hours after the loading dose and at 2 and 12 hours after delivery. Clinical signs of magnesium toxicity were carefully monitored. Maternal and neonatal outcome were evaluated. Significantly more women in the present study group reached the therapeutic level of serum magnesium at 2 hours (70% vs. 23%, p = 0.001) and at 4 hours (80% vs. 17%, p = 0.00) after the loading dose and at 2 hours (60% vs. 20%, p = 0.003) and at 12 hours (80% vs. 37%, p = 0.001) after delivery. No clinical magnesium toxicity was observed There were no significant differences in maternal and neonatal outcomes between the two groups. The maintenance dose of magnesium sulfate at 2 g/hour was more likely to attain the therapeutic level of serum magnesium when compared to 1 g/hour with no detectable difference in maternal and neonatal outcomes.
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)