Merck
Control of RhoA methylation by carboxylesterase I.
The Journal of biological chemistry (2013-05-10)
Ian Cushman, Stephanie M Cushman, Philip M Potter, Patrick J Casey
ABSTRACT
A number of proteins that play key roles in cell signaling are post-translationally modified by the prenylation pathway. The final step in this pathway is methylation of the carboxyl terminus of the prenylated protein by isoprenylcysteine carboxylmethyltransferase. Due to the impact of methylation on Rho function, we sought to determine if the process was reversible and hence could control Rho function in a dynamic fashion. Elevating isoprenylcysteine carboxylmethyltransferase activity in cells has profound effects on MDA-MB-231 cell morphology, implying the presence of a pool of unmethylated prenyl proteins in these cells under normal conditions. Using a knockdown approach, we identified a specific esterase, carboxylesterase 1, whose function had a clear impact not only on the methylation status of RhoA but also RhoA activation and cell morphology. These data provide compelling evidence that C-terminal modification of prenyl proteins, rather than being purely a constitutive process, can serve as a point of regulation of function for this important class of protein.
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg