Merck
  • Zn(2+) rather than Ca(2+) or Mg(2+) used as a cofactor in non-muscular actin from the oyster to control protein polymerization.
Zn(2+) rather than Ca(2+) or Mg(2+) used as a cofactor in non-muscular actin from the oyster to control protein polymerization.
Biochimica et biophysica acta (2013-05-04)
Xiaowei Yang, Chenyan Lv, Shengli Zhang, Guanghua Zhao, Changwei Ma
ABSTRACT
The major cytoskeletal protein of most cells is actin, which polymerizes to form actin filaments (F-actin). Each actin monomer (G-actin) contains a divalent alkaline earth metal ion (in vivo Mg(2+); in vitro usually Ca(2+)) as a cofactor that is crucial for protein polymerization. Prior to this study, however, whether or not other types of metal ions can play the same role as Mg(2+) or Ca(2+) in actins remains unknown. A new actin from the gills of oyster (AGO) was prepared and characterized by protein purification techniques, SDS- and native-PAGE, and LC-MS\MS for the first time. The property of this protein was studied by CD, fluorescence and UV/vis spectroscopy, laser light scattering, and TEM. AGO is a monomer with a MW of ~42kDa. AGO is unique among all known actins in that Zn(2+) is only a naturally binding metal in the protein, and that one native AGO molecule binds 8 zinc ions, which can be removed by EDTA treatment at pH7.2. The presence of zinc has a great effect on the secondary and tertiary structure of the protein. Correlated with such effect is that these zinc ions in native AGO facilitate protein polymerization, whereas removal of zinc ions from native AGO results in a loss of such polymerization property. The present work demonstrates that AGO is a novel zinc-binding protein with high capacity, and high selectivity. This work extends an understanding of the function of zinc and actin.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)