Merck
  • Hypoxia and cytoplasmic alkalinization upregulate growth hormone expression in lymphocytes.
Hypoxia and cytoplasmic alkalinization upregulate growth hormone expression in lymphocytes.
Cellular immunology (2013-05-04)
Douglas A Weigent
ABSTRACT
We report here that culture of lymphoid cells under hypoxic conditions showed an increase in both luciferase expression from a GH-promoter luciferase construct and the levels of lymphocyte GH. The effect was mimicked by treatment of cells with cobalt chloride consistent with a specific oxygen-sensing mechanism. We identified a putative hypoxia response element (HRE) in the GH promoter at the region -176 bp to -172 bp that contains a copy of the hypoxia-inducible factor-1 (Hif-1) binding motif (5'-ACGTG-3'). The results also showed that culture of primary rat spleen cells with different doses of TMA induced a dose-dependent increase in lymphocyte GH by Western blot analysis. Greater levels of GH are induced in T cell-enriched populations compared to B cell-enriched populations after treatment with CoCl(2) or TMA. Our results suggest that the stressful cellular conditions likely to occur at sites of inflammation or tumor growth may induce the synthesis of lymphocyte GH.
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt(II) chloride, 97%
Sigma-Aldrich
Cobalt(II) chloride, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Cobalt chloride 0.1 M solution