Merck
  • Homocysteine, folate, vitamin B-12, and 10-y incidence of age-related macular degeneration.
Homocysteine, folate, vitamin B-12, and 10-y incidence of age-related macular degeneration.
The American journal of clinical nutrition (2013-05-03)
Bamini Gopinath, Victoria M Flood, Elena Rochtchina, Jie Jin Wang, Paul Mitchell
ABSTRACT
Epidemiologic evidence of a relation between serum total homocysteine (tHcy), vitamin B-12, and folate and age-related macular degeneration (AMD) is inconsistent and unresolved. In this cohort study, we aimed to investigate associations between intakes and serum concentrations of folate and vitamin B-12 or serum tHcy and 10-y AMD incidence. Serum folate, vitamin B-12, and tHcy were determined from blood samples drawn in 1997-1999 from cohort members aged ≥55 y. AMD was assessed in 1760 survivors from retinal photographs taken in 2002-2004 and 2007-2009. Total intakes of folate and vitamin B-12 were assessed by using a food-frequency questionnaire. After adjustment for age, sex, current smoking, white blood cell count, and fish consumption, each 1-SD increase in serum tHcy was associated with increased risk of incident early and any AMD [ORs (95% CIs): 1.33 (1.09, 1.63) and 1.33 (1.11, 1.60), respectively]. Participants with a serum vitamin B-12 deficiency (<185 pmol/L) had higher risk of incident early and late AMD [ORs (95% CIs): 1.58 (1.06, 2.36) and 2.56 (1.38, 4.73), respectively]. Folate deficiency (<11 nmol/L) was associated with 75% and 89% increased risk of incident early and any AMD, respectively, 10 y later. Participants who reported supplementary vitamin B-12 intake had 47% reduced risk of incident any AMD (OR: 0.53; 95% CI: 0.33, 0.85). Elevated serum tHcy and folate and vitamin B-12 deficiencies predicted increased risk of incident AMD, which suggests a potential role for vitamin B-12 and folate in reducing AMD risk.
Sigma-Aldrich
L-Homocysteine, ≥98.0% (NT)
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, Vetec, reagent grade, ≥98%
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications