Merck
Genotoxicity and oxidative stress in gasoline station attendants.
Mutation research (2013-05-01)
Angela M Moro, Mariele F Charão, Natália Brucker, Juliano Durgante, Marília Baierle, Guilherme Bubols, Gabriela Goethel, Rafael Fracasso, Sabrina Nascimento, Rachel Bulcão, Bruna Gauer, Anelise Barth, Guilherme Bochi, Rafael Moresco, Adriana Gioda, Mirian Salvador, Sandra Farsky, Solange C Garcia
ABSTRACT
We evaluated genotoxic effects of exposure to low levels of benzene, a class I human carcinogen, among gasoline station attendants (GSA). Oxidative stress and the protective effects of antioxidants on DNA damage were also analyzed. Although exposures were below ACGIH (American Conference of Governmental Industrial Hygienists) limits, the GSA group presented higher DNA damage indices and micronucleus frequencies, increased oxidative protein damage, and decreased antioxidant capacity relative to the control group. Duration of benzene exposure was correlated with DNA and protein damage. The biomarkers evaluated in this work may provide early signals of damage in subjects occupationally exposed to benzene.
Product Description
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%