Merck
  • The use of hydrophilic poly(N,N-dimethylacrylamide) for promoting engulfment of magnetic gamma-Fe2O3 nanoparticles by mammalian cells.
The use of hydrophilic poly(N,N-dimethylacrylamide) for promoting engulfment of magnetic gamma-Fe2O3 nanoparticles by mammalian cells.
Journal of biomedical nanotechnology (2013-04-30)
Beata Anna Zasońska, Nataliya Boiko, Daniel Horák, Olga Klyuchivska, Hana Macková, Milan J Benes, Michal Babic, Miroslava Trchová, Jirina Hromádková, Rostyslav Stoika
ABSTRACT
gamma-Fe2O3 nanoparticles obtained by coprecipitation of Fe(II) and Fe(III) chlorides with a base and subsequent oxidation were coated with a shell of hydrophilic biocompatible poly(N,N-dimethylacrylamide) (PDMAAm). Various initiators were attached to the iron oxide surface to enable the use of the "grafting-from" approach for immobilization of PDMAAm. They included 2,2'-azobis(2-methylpropanimidamide) dihydrochloride (AMPA), 2,2'-azobis(N-hydroxy-2-methylpropanimidamide) dihydrochloride (ABHA) and 4-cyano-4-{[1-cyano-3-(N-hydroxycarbamoyl)-1-methylpropyl]azo}pentanoic acid (CCHPA). Engulfment of PDMAAm-coated y-Fe2O3 nanoparticles by murine J774.2 macrophages was investigated. Only some nanoparticles were engulfed by the macrophages. PDMAAm-AMPA-gamma-Fe2O3 and PDMAAm-ABHA-y-Fe2O3 nanoparticles were rapidly engulfed by the cells. In contrast, neat y-Fe2O3 and PDMAAm-CCHPA-gamma-Fe2O3 particles induced formation of transparent vacuoles indicating toxicity of the particles. Thus, PDMAAm-coated AMPA- and ABHA-gamma-Fe2O3 nanoparticles can be recommended as non-toxic labels for mammalian cells.
Sigma-Aldrich
Iron(III) oxide, nanopowder, <50 nm particle size (BET)
Sigma-Aldrich
Iron(III) oxide, ≥99.995% trace metals basis
Sigma-Aldrich
Iron(III) oxide, powder, <5 μm, ≥96%
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol