Merck
  • Enhanced delivery system of flutamide loaded chitosan-dextran sulphate nanoparticles for prostate cancer.
Enhanced delivery system of flutamide loaded chitosan-dextran sulphate nanoparticles for prostate cancer.
Journal of biomedical nanotechnology (2013-04-30)
A Anitha, Saji Uthaman, Shantikumar V Nair, R Jayakumar, Vinoth-Kumar Lakshmanan
ABSTRACT
In the current work, a sustained drug delivery system of flutamide (FLT) was developed using chitosan (CS) and dextran sulphate (DS) nanoparticles and were characterized using different techniques. The prepared nanoparticles showed a size of 80-120 nm with an entrapment efficiency of 55 +/- 6.95%. In addition, blood compatibility, in vitro cytotoxicity, drug release and cellular uptake studies were also carried out. The drug release studies showed a sustained and pH dependent release pattern as a result, after 120 h about 66% drug release occurred at pH 7.4 and 78% release occurred in acidic pH. MTT (3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide) and LDH (lactate dehydrogenase) experiments proved the preferential toxicity of drug loaded nanoparticles towards prostate cancer cells (PC3) unlike in normal cells, mouse fibroblast cells (L929). The cell death mechanism of drug loaded nanoparticles for a concentration of 50 and 75 nM showed 28 +/- 2 and 35.2 +/- 4% apoptosis in samples treated with the PC3 cells after 24 h. Fluorescent microscopic imaging and flow cytometry confirmed the preferential uptake of the nanoparticles (NPs) in the prostate cancer cells (PC3) unlike in normal (L929) cells. Hence the developed FLT loaded CS-DS NPs could be used as a promising system for controlled delivery in prostate cancer.
Sigma-Aldrich
Flutamide
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Supelco
Dextran, analytical standard, Set Mp 1,000-400,000
Supelco
Dextran, analytical standard, 670,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 670,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 12,000
Sigma-Aldrich
Dextran, enzymatic synth.
Supelco
Dextran, analytical standard, 410,000
Supelco
Dextran, analytical standard, 150,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Supelco
Dextran, analytical standard, 5,000
Supelco
Dextran, analytical standard, 25,000
Supelco
Dextran, analytical standard, 1,000
Supelco
Dextran, analytical standard, 270,000
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, medium molecular weight
Supelco
Dextran, analytical standard, 80,000
Supelco
Dextran, analytical standard, 50,000