Merck
  • Formulation, in vitro drug release study and anticancer activity of 5-fluorouracil loaded gellan gum microbeads.
Formulation, in vitro drug release study and anticancer activity of 5-fluorouracil loaded gellan gum microbeads.
Acta poloniae pharmaceutica (2013-04-25)
S K Sahoo, S K Sahoo, A Behera, S V Patil, S K Panda
ABSTRACT
5-Fluorouracil loaded calcium-zinc-gellan and calcium-zinc-gellan-ethyl cellulose microbeads were successfully prepared by simple ionotropic gelation and oil in water ionotropic gelation technique, respectively. Prepared microbeads were characterized by scanning electron microscopy, Fourier transform infrared spectroscopy and evaluated for particle size, drug content, encapsulation efficiency, drug release and cell cytotoxicity study. Microbeads formed were spherical with rough surface. As concentration of gellan and ethyl cellulose has increased encapsulation efficiency, particle size and sustained drug release effect also increased. The release of 5-fluorouracil from microbeads has followed Hixson Crowell model suggesting the mechanism of drug release as dissolution controlled. Cytotoxicity analysis on HT-29 human colon cancer cell lines indicated that 5-FU loaded gellan gum/gellan in combination with ethyl cellulose microbeads leads to sustained releases of drug and thus delayed apoptosis over a long period of time. The formulation with drug:gellan:ethyl cellulose ratio 2.5:7.5:1 was found to be more effectual in terms of sustained drug release activity in addition to anti-cancer activity.
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Zinc sulfate heptahydrate, suitable for plant cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, meets USP testing specifications
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Zinc sulfate heptahydrate, BioUltra, for molecular biology, 2.0 M in H2O
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Sigma-Aldrich
Zinc sulfate heptahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, Vetec, reagent grade, 99%
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Zinc sulfate solution, 0.3 N
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis