Merck
  • In vitro reconstitution of complexes between pro-matrix metalloproteinase-9 and the proteoglycans serglycin and versican.
In vitro reconstitution of complexes between pro-matrix metalloproteinase-9 and the proteoglycans serglycin and versican.
The FEBS journal (2013-04-23)
Nabin Malla, Eli Berg, Achilleas D Theocharis, Gunbjørg Svineng, Lars Uhlin-Hansen, Jan-Olof Winberg
ABSTRACT
Previously, we have shown that a proportion of the matrix metalloproteinase-9 (MMP-9) synthesized by the macrophage cell line THP-1 binds to a chondroitin sulfate proteoglycan (CSPG) core protein to form a reduction-sensitive heteromer. It was also shown that the hemopexin-like (PEX) domain and the fibronectin-like (FnII) module in the enzyme are involved in heteromer formation. In this paper, we show that reduction-sensitive and SDS-stable heteromers may be reconstituted in vitro by mixing proMMP-9 with either serglycin, versican or CSPGs isolated from various monocytic cell lines. In addition, a strong but SDS-soluble proMMP-9·CSPG heteromer was formed. The two macromolecules in the SDS-stable reduction-sensitive heteromers were not linked together by disulfide bonds. As for the heteromer isolated from THP-1 cells, in vitro reconstituted SDS-stable and SDS-soluble heteromers showed weaker binding to gelatin than the proMMP-9 monomer. Furthermore, gelatin inhibited in vitro reconstitution of the heteromers, showing that the FnII module is involved in the complex formation. Tissue inhibitor of metalloproteinase (TIMP)-1 was not be detected in the proMMP-9·CSPG complexes. However, the presence of TIMP-1 inhibited formation of the SDS-soluble heteromer, but not the SDS-stable reduction-sensitive heteromer. This indicates that different regions in the PEX domain are involved formation of these heteromers.
Product Description
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Triton X-102
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Cystine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Cystine
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free