Merck
  • Lectin-like oxidized LDL receptor-1 is palmitoylated and internalizes ligands via caveolae/raft-dependent endocytosis.
Lectin-like oxidized LDL receptor-1 is palmitoylated and internalizes ligands via caveolae/raft-dependent endocytosis.
Biochemical and biophysical research communications (2013-04-16)
Miyuki Kumano-Kuramochi, Qiuhong Xie, Shoko Kajiwara, Shiro Komba, Takashi Minowa, Sachiko Machida
ABSTRACT
Lectin-like oxidized low-density lipoprotein (LDL) receptor-1 (LOX-1) is an endothelial scavenger receptor that is important for oxidized low-density lipoprotein uptake. LOX-1 functions as an oligomer; however, little is known about the oligomeric complex and ligand processing after recognition by LOX-1. Here, we found that LOX-1 recognized and internalized ligands through the caveolae/raft-dependent endocytosis pathway in human coronary artery endothelial cells. Furthermore, we demonstrated that LOX-1 was palmitoylated and that both cysteine 36 and cysteine 46 were necessary for the recruitment of LOX-1 into raft microdomains and for its ligand uptake ability.
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Supelco
Genistein, analytical standard
Sigma-Aldrich
Nystatin, ≥4,400 USP units/mg
Sigma-Aldrich
Nystatin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nystatin, powder, γ-irradiated, BioXtra, suitable for cell culture
Nystatin, European Pharmacopoeia (EP) Reference Standard