Merck
Illustrative cases of unorthodox tinnitus management.
Noise & health (2013-04-11)
Kenneth H Brookler
ABSTRACT
Bone remodeling disorders have been identified as potential candidates for the etiology underlying some forms of otologic disease. Here, four clinical cases are presented to demonstrate a novel approach to tinnitus management that focuses on two fundamental etiologic bases. First, is the effect of bone remodeling abnormalities of the otic capsule and second, the effect of blood sugar and insulin abnormalities upon the function of the inner ear and self-reports of tinnitus. This report presents the effect of treatment directed at bone remodeling abnormalities of the otic capsule and abnormalities of blood sugar and insulin. The treatment effects led to a substantial reduction in subjective tinnitus. These findings suggest a potential convergence between bone pathophysiology and some forms of tinnitus.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%