Merck
  • Cobryketone derived from vitamin B12 via palladium-catalyzed cleavage of the sp3-sp3 carbon-carbon bond.
Cobryketone derived from vitamin B12 via palladium-catalyzed cleavage of the sp3-sp3 carbon-carbon bond.
The Journal of organic chemistry (2013-04-03)
Sylwester Kurcoń, Keith ó Proinsias, Dorota Gryko
ABSTRACT
Heptamethyl cobyrinate was transformed into hexamethyl 8-nor-cobyrinate. The crucial step involved the synthesis of new, vitamin B12 derived cobryketone via palladium-catalyzed cleavage of the sp(3)-sp(3) carbon-carbon bond with the liberation of the ketone. The replacement of sp(3) carbon atom with sp(2) (C═O) at the 8-position produces a bathochromic shift of all absorption bands and makes α and β bands equal as a consequence of the expansion of the existing conjugated system of double bonds.
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material