Merck
  • Iodine(III)-mediated umpolung of bromide salts for the ethoxybromination of enamides.
Iodine(III)-mediated umpolung of bromide salts for the ethoxybromination of enamides.
Organic letters (2013-04-02)
Sophie Nocquet-Thibault, Pascal Retailleau, Kevin Cariou, Robert H Dodd
ABSTRACT
Using (diacetoxyiodo)benzene in conjunction with simple bromide salts in ethanol allows the regioselective ethoxybromination of a wide range of enamides, thus yielding highly versatile α-bromo hemiaminals, which can then be engaged in a broad array of transformations.
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis