Merck
  • Growth and hematology of juvenile pacu Piaractus mesopotamicus (Holmberg 1887) fed with increasing levels of vitamin E (DL-α-tocopheryl acetate).
Growth and hematology of juvenile pacu Piaractus mesopotamicus (Holmberg 1887) fed with increasing levels of vitamin E (DL-α-tocopheryl acetate).
Anais da Academia Brasileira de Ciencias (2013-03-30)
Ricardo Y Sado, Alvaro J A Bicudo, José E P Cyrino
ABSTRACT
Intensive fish production systems are characterized by 100% artificial feeding, so any dietary imbalances or deficiencies may lead to diseases outbreaks and economic losses. This study was set out to determine the effects of increasing levels of dietary vitamin E on growth and hematology of juvenile pacu. Fishes were fed for 90 days, twice a day until apparent satiation with semi-purified diets containing 0.0; 25; 50; 150; 300 or 600 mg.kg-1 diet DL-α-tocopheryl acetate in a completely randomized design trial (n=4); biometrical and hematological data were collected and analyzed. Fishes fed with vit E diet (150 mg.kg-1) showed higher (p<0.05) weight gain and specific growth. Hematocrit, erythroblast number and total plasma protein were increased (p<0.05) in fishes fed diet with no vit E diet. Vitamin E supplementation in artificial diets for pacu is essential for growth and maintenance of normal erythropoiesis.
Product Description
Supelco
Tocopheryl Acetate, a, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
DL-alpha-Tocopherol acetate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
DL-α-Tocopherol acetate, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
DL-α-Tocopherol acetate, ≥96% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Pricing and availability is not currently available.
α-Tocopherol acetate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
All-rac-alpha-tocopheryl acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.