Merck
  • Magnesium sulphate for prevention of eclampsia: are intramuscular and intravenous regimens equivalent? A population pharmacokinetic study.
Magnesium sulphate for prevention of eclampsia: are intramuscular and intravenous regimens equivalent? A population pharmacokinetic study.
BJOG : an international journal of obstetrics and gynaecology (2013-03-28)
D H Salinger, S Mundle, A Regi, H Bracken, B Winikoff, P Vicini, T Easterling
ABSTRACT
To compare magnesium sulphate concentrations achieved by intramuscular and intravenous regimens used for the prevention of eclampsia. Low-resource obstetric hospitals in Nagpur and Vellore, India. Pregnant women at risk for eclampsia due to hypertensive disease. A pharmacokinetic study was performed as part of a randomised trial that enrolled 300 women comparing intramuscular and intravenous maintenance regimens of magnesium dosing. Data from 258 enrolled women were analysed in the pharmacokinetic study. A single sample was drawn per woman with the expectation of using samples in a pooled data analysis. Pharmacokinetic parameters of magnesium distribution and clearance. Magnesium clearance was estimated to be 48.1 dl/hour, volume of distribution to be 156 dl and intramuscular bioavailability to be 86.2%. The intramuscular regimen produced higher initial serum concentrations, consistent with a substantially larger loading dose. At steady state, magnesium concentrations in the intramuscular and intravenous groups were comparable. With either regimen, a substantial number of women would be expected to have serum concentrations lower than those generally held to be therapeutic. Clinical implications were that a larger loading dose for the intravenous regimen should be considered; where feasible, individualised dosing of magnesium sulphate would reduce the variability in serum concentrations and might result in more women with clinically effective magnesium concentrations; and lower dose magnesium sulphate regimens should be considered with caution.
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)