Merck
  • [The combined therapy with valdoxan and amitriptyline of treatment resistant somatized depression].
[The combined therapy with valdoxan and amitriptyline of treatment resistant somatized depression].
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova (2013-03-27)
V S Sobennikov, M L Prokop'eva
ABSTRACT
The combined treatment with valdoxan and amitriptyline has been suggested as one of the possible variants to overcome treatment resistance in somatized depression. A study design was based on the rhythmological model of depression which clinically characterized by altered sleep-awakeness cycles. According to this notion, 36 patients were divided into two groups by the presence (20 patients) or absence (16 patients) of severe sleep disorders. The combined treatment was more effective in the first group of patients with insomnia which was an indicator of altered sleep-awakeness cycles.
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
Agomelatine, ≥98% (HPLC)