Merck
  • Cross-talk between constitutive androstane receptor and hypoxia-inducible factor in the regulation of gene expression.
Cross-talk between constitutive androstane receptor and hypoxia-inducible factor in the regulation of gene expression.
Toxicology letters (2013-03-27)
Ryota Shizu, Sawako Shindo, Takemi Yoshida, Satoshi Numazawa
ABSTRACT
Hypoxia inducible factor (HIF) and 5'-AMP-activated protein kinase are often activated under similar physiological conditions. Constitutive androstane receptor (CAR) translocates into the nucleus in accordance with 5'-AMP-activated protein kinase and thus confers transactivation. The aim of the present study was to investigate a possible link between CAR and HIFα. Phenobarbital (PB), a typical CAR activator, increased the gene expression of HIF-target genes in the livers of mice, including erythropoietin, heme oxygenase-1 and vascular endothelial growth factor-a. PB induced an accumulation of nuclear HIF-1α and an increase in the HIF-responsive element-mediated transactivation in HepG2 cells. Cobalt chloride, a typical HIF activator, induced the gene expression of CAR-target genes, including cyp2b9 and cyp2b10, an accumulation of nuclear CAR and an increase in the PB-responsive enhancer module-mediated transactivation in the mouse liver. Immunoprecipitation-immunoblot and chromatin immunoprecipitation analyses suggest that CAR binds to the PB-responsive enhancer module with HIF-1α in the liver of untreated mice and that the complex dissociates upon PB treatment. Taken together these results suggest that CAR and HIF-α interact and reciprocally modulate the functions of each other.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt(II) chloride, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
Cobalt(II) chloride, 97%
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Phenobarbital sodium salt
Sigma-Aldrich
Phenobarbital
Sigma-Aldrich
Cobalt chloride 0.1 M solution