Merck
  • Advanced age as a risk factor for folate-associated functional cobalamin deficiency.
Advanced age as a risk factor for folate-associated functional cobalamin deficiency.
Journal of the American Geriatrics Society (2013-03-26)
Lawrence R Solomon
ABSTRACT
To determine whether high serum folate levels contribute to metabolite changes in elderly subjects with normal cobalamin levels. Case series. Outpatient clinic at a university-based staff model health maintenance organization. Two hundred thirty-three ambulatory individuals without diabetes mellitus with normal renal function and normal cobalamin levels evaluated for cobalamin deficiency. Cobalamin, serum folate, methylmalonic acid (MMA), and homocysteine. Older individuals (≥60) with low-normal cobalamin levels (201-300 pg/mL) had higher MMA and lower homocysteine levels when serum folate levels were high (>20 ng/mL) than when serum folate levels were normal (P < .02), but serum folate levels within the normal range were not a determinant of either metabolite. In younger subjects with low-normal cobalamin levels, high serum folate levels were not associated with significant differences in either metabolite. At mid-normal cobalamin levels (301-600 pg/mL), high serum folate levels were associated with lower homocysteine levels in older adults (P < .001) but not with differences in MMA in either age group. Cobalamin therapy decreased or normalized MMA and homocysteine in 89% or more of participants even at pretherapy cobalamin levels greater than 600 pg/mL. High serum folate levels are associated with higher MMA levels when cobalamin levels are low-normal, and this effect is age dependent, not progressive within the normal serum folate range (suggesting a threshold effect), and reversed by cobalamin therapy. Because MMA may be neurotoxic, these findings suggest caution in the use of folic acid supplements in elderly adults.
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, Vetec, reagent grade, ≥98%
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Methylmalonic acid, 99%