Merck
  • Huge increase of therapeutic window at a bioactive silver/titania nanocomposite coating surface compared to solution.
Huge increase of therapeutic window at a bioactive silver/titania nanocomposite coating surface compared to solution.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Tomislav Hrkac, Claudia Röhl, Rainer Podschun, Vladimir Zaporojtchenko, Thomas Strunskus, Heike Papavlassopoulos, Dieter Garbe-Schönberg, Franz Faupel
ABSTRACT
A silver containing coating used in the human body, e.g., on an implant should be both effectively antimicrobial and non-cytotoxic to human cells. It is generally believed that the biologic effect originates from silver ions released from the coating. Nanocomposites with well controlled Ag filling factor were prepared by co-sputtering, and the silver surface concentration and the silver release were determined by XPS and ICP-MS, respectively. Here we show that only a small therapeutic window exists for dissolved silver but the therapeutic window is largely increased at the surface. While the toxicity observed for mammalian cells in contact with the bioactive Ag/TiO2 nanocomposite surface and for silver ions in solution is rather similar the antimicrobial activity is drastically enhanced at the surface. A model is proposed to explain the strong increase of the antimicrobial activity at the surface. The present results not only question well-established tests for antimicrobial activity but they are also important for the design of antimicrobial coatings, e.g., for biomedical devices.
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh