Merck
  • Role of tentacles and protein loading on pore accessibility and mass transfer in cation exchange materials for proteins.
Role of tentacles and protein loading on pore accessibility and mass transfer in cation exchange materials for proteins.
Journal of chromatography. A (2013-03-16)
Helen Thomas, Bertrand Coquebert de Neuville, Giuseppe Storti, Massimo Morbidelli, Matthias Joehnck, Michael Schulte
ABSTRACT
In protein chromatography, the size of the protein determines which fraction of pores it can access within a resin and at which rate of diffusion. Moreover, in the presence of grafted polymers like in advanced materials, adsorbed proteins and electrolytes complicate the interaction pore-protein. In this study, we evaluated in a comparative way the behavior of Fractogel EMD SO3 (M) and (S), "tentacle"-type, strong cation exchangers, as well as a reference material without tentacles, all of which are commonly used for protein purification. ISEC experiments were carried out with a set of Dextran tracers of largely different molecular size covering the typical range of protein sizes. Experimental values of porosity (internal and external to the particles) as well as of pore diffusion coefficients have been measured at different NaCl concentrations and under protein loading. These results provide useful insights into the complex interplay among mentioned factors: first, the presence of tentacles induces size exclusion selectivity in the materials; second, the salt induces conformational changes of the tentacles, leading to porosities larger than expected in tentacle materials; third, protein adsorption mainly leads to a reduction of porosity due to pore space occupied by the protein and to a decrease of pore diffusion coefficient.
Sigma-Aldrich
Kollidon® 25
Povidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Povidone, meets USP testing specifications
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000