Merck
  • Kinetic study of the 7-endo selective radical cyclization of N-tert-butyl-o-bromobenzylmethacryl amides: kinetic investigation of the cyclization and 1,7-hydrogen transfer of aromatic radicals.
Kinetic study of the 7-endo selective radical cyclization of N-tert-butyl-o-bromobenzylmethacryl amides: kinetic investigation of the cyclization and 1,7-hydrogen transfer of aromatic radicals.
The Journal of organic chemistry (2013-03-16)
Akio Kamimura, Tomoko Kotake, Yuriko Ishihara, Masahiro So, Takahiro Hayashi
ABSTRACT
A kinetic investigation of the radical cyclization of N-tert-butyl-o-bromobenzylmethacryl amides to give 2-benzazepines via 7-endo selective cyclization was undertaken. The aryl radical generated from the amide precursor by treatment with Bu3SnH gave the three compounds, which are a 7-endo cyclized adduct, a 6-exocyclized adduct, and a reduced product. The cyclization reactions under various Bu3SnH concentrations were traced by GC analysis. The 7-endo/6-exo selectivity was constant irrespective of variation in Bu3SnH concentration. These results revealed that regioselectivity is controlled in a kinetic manner and that there is no possibility of a neophyl rearrangement. The use of Bu3SnD revealed that 1,7-hydrogen transfer, in which an aryl radical abstracts a hydrogen atom from the methallylic methyl group, occurs during the reaction. Hydrogen abstraction from toluene, the reaction solvent, was also observed. The 1,7-transfer rate depended on the Bu3SnX (X = H or D), and the reaction kinetics was examined. The k(H)/k(D) value for the hydrogen abstraction of aryl radical from Bu3SnX (X = H or D) was estimated using 4-bromoanisol. The utilization of these values revealed the overall reaction kinetics and relative rates for the cyclization and reduction by Bu3SnX (X = H or D). Kinetic parameters for hydrogen abstraction from toluene by aryl radicals were also estimated.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%