Merck
  • How a simple "clicked" PEGylated 1,2,3-triazole ligand stabilizes gold nanoparticles for multiple usage.
How a simple "clicked" PEGylated 1,2,3-triazole ligand stabilizes gold nanoparticles for multiple usage.
Chemical communications (Cambridge, England) (2013-03-14)
Pengxiang Zhao, Na Li, Lionel Salmon, Na Liu, Jaime Ruiz, Didier Astruc
ABSTRACT
"Click" chemistry now offers access to a great variety of triazoles, and the first example of a strategy to stabilize gold nanoparticles (AuNPs) with a new 1,2,3-triazole-mPEG ligand is developed here together with preliminary examples of possible applications.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350