Merck
  • Dynamic bone imaging with 99mTc-labeled diphosphonates and 18F-NaF: mechanisms and applications.
Dynamic bone imaging with 99mTc-labeled diphosphonates and 18F-NaF: mechanisms and applications.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2013-03-14)
Ka Kit Wong, Morand Piert
ABSTRACT
Dynamic bone scanning with (99m)Tc-labeled diphosphonates and (18)F-labeled sodium fluoride provides functional information sensitive for subtle changes in bone turnover and perfusion, which assists the clinical management of numerous osseous pathologies. This article reviews the mechanisms of uptake of (99m)Tc-labeled diphosphonates and (18)F-sodium fluoride and discusses and compares the performance of these bone-seeking radiotracers for clinical and research applications, using dynamic and multiple-time-point imaging protocols and quantitative techniques.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%