Merck
Clinical implications of CES2 RNA expression in neuroblastoma.
Journal of pediatric surgery (2013-03-14)
Keiichi Uchida, Kohei Otake, Koji Tanaka, Kiyoshi Hashimoto, Susumu Saigusa, Kohei Matsushita, Yuhki Koike, Mikihiro Inoue, Motoko Ueeda, Yoshinaga Okugawa, Yasuhiro Inoue, Yasuhiko Mohri, Masato Kusunoki
ABSTRACT
Human carboxylesterase 2 (CES2) is the key enzyme for metabolic activation of irinotecan (CPT-11). The aim was to evaluate the clinical implications of CES2 RNA expression in neuroblastoma cells. CES2 RNA expression was determined by real-time reverse transcription-polymerase chain reaction in five neuroblastoma cell lines and 42 clinical samples of untreated neuroblastoma. Sensitivity to CPT-11 was assessed by WST-8 colorimetric assays. Induction of apoptosis was evaluated by flow cytometry after CPT-11 exposure. Protein expression of CES2 was evaluated by Western blotting analysis. CES2 RNA expression in clinical samples was investigated for its associations with the clinicopathological characteristics. CES2 RNA expression was observed in neuroblastoma cells, and its expression in neuroblastoma cell lines was positively correlated with sensitivity to CPT-11 and apoptosis after CPT-11 exposure in vitro. CES2 RNA expression was correlated with the protein levels of CES2 in vitro. CES2 RNA expression was significantly higher in patients with a characteristic related to advanced disease. Our results suggest the potential of clinical application of CPT-11 in neuroblastoma treatment for patients with advanced disease.
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg