Merck
Ultrasonic attenuation and speed of sound of cornstarch suspensions.
The Journal of the Acoustical Society of America (2013-03-08)
Benjamin L Johnson, Mark R Holland, James G Miller, Jonathan I Katz
ABSTRACT
The goal of this study is to contribute to the physics underlying the material properties of suspensions that exhibit shear thickening through the ultrasonic characterization of suspensions of cornstarch in a density-matched solution. Ultrasonic measurements at frequencies in the range of 4 to 8 MHz of the speed of sound and the frequency-dependent attenuation properties are reported for concentrations of cornstarch in a density-matched aqueous (cesium chloride brine) suspension, ranging up to 40% cornstarch. The speed of sound is found to range from 1483 ± 10 m/s in pure brine to 1765 ± 9 m/s in the 40% cornstarch suspension. The bulk modulus of a granule of cornstarch is inferred to be 1.2(± 0.1) × 10(10) Pa. The attenuation coefficient at 5 MHz increases from essentially zero in brine to 12.0 ± 1.2 dB/cm at 40% cornstarch.
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Cesium, ingot, ≥99.95% trace metals basis
Sigma-Aldrich
Cesium chloride, ReagentPlus®, 99.9%
Sigma-Aldrich
Cesium chloride, puriss. p.a., ≥99.5%
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥98%
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Starch from rice